Learn More
New Pig CleanMax™ Cleanroom Frock
Description
- High-Density, Low-Linting Material: Made from a high-density polyethylene film that is laminated to a continuous filament polypropylene nonwoven, the fabric is designed to provide an excellent barrier to particles while generating minimal lint and fibers.
- Strong and Durable: The laminated construction provides enhanced strength and abrasion resistance, making the frock durable enough to withstand the rigors of use in a controlled environment without compromising its protective qualities.
- Excellent Liquid Barrier: The poly-laminated fabric offers a superior barrier to liquid splashes and spills, providing a high level of protection from water-based liquids and certain chemicals.
- Comfortable and Breathable: While offering a strong barrier, the material is also designed to be breathable, allowing for the passage of moisture vapor and helping to reduce heat stress for the wearer.
- Integrated Features for a Secure Fit: The frock is designed with a front zipper closure with a storm flap and elastic wrists, which helps to create a secure fit and prevent the ingress of contaminants.
- Economical Choice: The CleanMax™ product line is often positioned as an economical alternative to more expensive cleanroom garments, providing a high level of protection at a lower cost for applications that require a balance of performance and value.
Specifications
Specifications
| Size | 5X-Large |