missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clean-Blot™ IP Detection Reagent (HRP)

Catalog No. PI21230
Change view
Click to view available options
Quantity:
2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21230 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21230 Supplier Thermo Scientific™ Supplier No. 21230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Avoid interference from denatured IP antibody fragments in Western blots of immunoprecipitated proteins with these HRP and AP conjugates.

Thermo Scientific™ Clean-Blot IP Detection Reagents are HRP and AP conjugates that are optimized for post-immunoprecipitation Western blot detection of primary antibodies without interference from denatured IP antibody fragments.

Clean-Blot IP Reagents allow trouble-free Western blot detection of target proteins following IP assays (immunoprecipitation). They function by specifically binding to functional primary antibodies (whole IgG) without also binding to fragments of the IP antibodies, which usually accompany the immunoprecipitated protein through electrophoresis and membrane transfer. The Clean-Blot IP Reagents and HRP Kit eliminate detection-interference from both heavy-chain (approx. 50kDa) and light-chain (25kDa) IgG-fragments of antibodies used for the initial immunoprecipitation assay. Clean-Blot IP products include detection reagents with alkaline phosphatase conjugate or horseradish peroxidase conjugate and the complete Clean-Blot IP Detection Kit (HRP) for chemiluminescent substrate development.

Highlights:

  • Universal – bind and detect most species, IgG subclasses and isotypes of primary antibodies that are commonly used for Western blotting

  • Compatible – effective with IP assays performed using Protein A, Protein G, or anti-IgG agarose beads and any blocking buffer

  • Cost-effective – eliminates the cost and extra work associated with covalently immobilizing IP antibodies as a means of overcoming Western blot interference

  • Flexible – available in alkaline phosphate (AP) and horseradish peroxidase (HRP) reagents for detection with chemiluminescent, fluorescent or colorimetric substrates

  • Easy to use – no need to change the Western blotting protocol; simply replace conventional secondary HRP conjugate with the Clean-Blot IP Detection Reagent

  • Unobstructed detection – clear Western blot results for immmunoprecipitation assays without significant interference from denatured IgG bands

Specifications

Description Clean-Blot IP Detection Reagent (HRP)
Product Type IP Detection Reagent (HRP)
Detection Method Chemiluminescence, Fluorescence, Colorimetric
Form Liquid
For Use With (Application) Western Blot
Product Line Pierce
Quantity 2.5 mL
Conjugate HRP
Reactivity All species
Content And Storage Store at 4°C.
How does the Clean-Blot IP Detection Kit work?

This assay uses a HRP conjugate or AP conjugate that binds to native IgG from various host species, allowing clear, specific Western blot detection from IP experiments without interference from denatured IgG. It is used simply as a substitute for secondary antibodies.

For Research Use Only. Not for use in diagnostic procedures.