missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ ArcGuard™ Class 0 Rubber Voltage Gloves Premium Kit

Catalog No. p-7220141
Change view
Click to view available options
Size:
10
11
12
8
9
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011990 8
170011991 9
170011992 10
170011993 11
170011994 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 170011990 Supplier National Safety Apparel™ Supplier No. KITGC008AG
Only null left

Premium kit includes a pair of electrical insulating rubber gloves, fire-resistant knit gloves, leather glove protectors, and a glove bag. Class 00 insulating gloves will protect workers from up to 500 V.

  • Electrical insulating gloves protect against electrical shock and other arc flash hazards
  • Wear with included leather glove protectors for additional resistance against mechanical hazards
  • FR knit gloves stretch for dexterity and can be worn for another layer of protection
  • Compliant with ANSI/ASTM D120 and NFPA 70E standards
  • Class 0 gloves offer protection from up to 1000 V
  • Gloves should be used within one year of marked test date
  • When not in use, lay flat in the included protective bag
  • Store away from direct sunlight, high heat, and humidity

Specifications

Certifications/Compliance ANSI/ASTM D120, Class 0, Type I, NFPA 70E, ASTM F1506, OSHA 1910.269
Applications Industrial
Color Black
Material Rubber, Leather
Product Type Electrical Insulating Gloves Kit
Size 8
For Use With (Application) Electrical Utility Work, Shock Protection
Glove Cuff Style Rolled
Series KITGC0BAG
Voltage 1000 V