missing translation for 'onlineSavingsMsg'
Learn More

Clarinex™ Desloratidine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012277
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012277 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012277 Supplier Merck Emd Millipore Supplier No. PHR16801G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Desloratadine is one of the orally active, highly selective peripheral H1 receptor antagonists that does not exert any significant effect on the central and autonomic nervous systems. The potency of desloratadine is more than that of loratadine and it exhibits substantial overall pharmacological profile. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 100643-71-8
Product Line Clarinex
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C19H19 ClN2
MDL Number MFCD00871949
Synonym Desloratadine; 8-Chloro-6,11-dihydro-11-(4-piperidinylidene)-5 H- benzo[5,6 ]cyclohepta[1,2,b]pyridine; Clarinex(R); NSC 675447
Type Certified Reference Material
Quantity 1 g
Formula Weight 310.82
Grade Certified Reference Material

Clarinex is a registered trademark of Schering Corp.