Learn More
Thermo Scientific™ Clamps for CO2 Resistant Shaker
Achieve maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
Supplier:  Thermo Scientific™ 88881114
 View more versions of this product
Catalog No. 88-881-114
Only null left
Explore available promotions
Description
Description
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
  • Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Specifications
Flask Clamp
5000 mL
CO2 Resistant Shaker
1
Product Suggestions
Product Suggestions