Learn More
Brady™ Clamp-On Circuit Breaker Lockout
Description
Experience reliable lockout safety and effortless installation with the Clamp-On Circuit Breaker Lockout. Simply twist the proprietary thumbwheel design for tool-free application.
- Fits a wide range of single-pole and internal-trip multi-pole breakers. Detachable cleat expands the range of applicable breakers.
- Made from fiberglass-reinforced nylon for durability in demanding environments.
- Tamper-resistant features help prevent unauthorized removal.
- Convenient size for easy transport and storage.
- Accepts padlock with shackle diameter up to 0.28 in.
Specifications
Specifications
| Product Type | Circuit Breaker Lockout |
| Material | Polycarbonate |
| Description | Clamp-on Circuit Breaker |
| For Use With (Equipment) | 480/600 V Circuit Breaker |
| Includes | Attachable Cleat for Circuit Breaker Lockout Device |
| Quantity | 1 Ea. |