Learn More
Fisherbrand™ Clamp, Chain
Chain Clamp holds large round or irregular shaped objects firmly, yet gently to lab-frames and rods
Supplier:  Fisherbrand™ CC500170
Numéro de catalogue. 01257565
Il en reste null
Description
Description
  • Quick and secure slip-on chain connection with large, easy-to-turn adjusting knob
  • Extension arm allows user to vary distance from the frame
  • With electro-polished stainless steel chain
Spécifications
Spécifications
Autoclavable
Zinc Alloy
Chain, Knob
12 mm
7.9 in.
5.7 in.
145 mm
170 mm
6.7 in.
V
Chain
Chain Clamp
Up to 170 mm dia.
0.47 in.
200 mm
7.9 in.
200 mm
Standard
2 in.
50 mm
Silver
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS