Learn More
Chemglass Life Sciences Adapter, Claisen, 29/42 Joint Size, 175mm H x 105mm W
Supplier: Chemglass Inc CG102002
Adapter, Claisen, 29/42 Joint Size, 175mm H x 105mm W. Outer standard taper joints on both top arms and lower inner joint. All three joints are the same size.