missing translation for 'onlineSavingsMsg'
Learn More

Citronellol, 95%

Catalog No. AAA1901622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1901622 100 g
AAA1901636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1901622 Supplier Thermo Scientific Chemicals Supplier No. A1901622

CAS: 106-22-9 | C10H20O | 156.269 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-22-9
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00002935
InChI Key QMVPMAAFGQKVCJ-UHFFFAOYSA-N
Synonym citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol
PubChem CID 8842
ChEBI CHEBI:50462
IUPAC Name 3,7-dimethyloct-6-en-1-ol
SMILES CC(CCC=C(C)C)CCO

Specifications

Melting Point <-20°C
Density 0.858
Boiling Point 221°C to 222°C
Flash Point 98°C (208°F)
Odor Pleasant
Refractive Index 1.4575
Quantity 100 g
UN Number UN3082
Beilstein 1721507
Merck Index 14,2330
Formula Weight 156.27
Percent Purity 95%
Chemical Name or Material Citronellol
Show More Show Less
Hazard Category H227-H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 203-375-0
RTECSNumber RH3400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only