missing translation for 'onlineSavingsMsg'
Learn More

Citronellol, 95%

CAS: 106-22-9 | C10H20O | 156.27 g/mol

Supplier:  Thermo Scientific Chemicals 110461000

Quantity 100 mL
Packaging Glass bottle

 View more versions of this product

Catalog No. AC110461000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-22-9
156.27
QMVPMAAFGQKVCJ-UHFFFAOYSA-N
8842
3,7-dimethyloct-6-en-1-ol
C10H20O
MFCD00002935
citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol
CHEBI:50462
CC(CCC=C(C)C)CCO

Specifications

Citronellol
106-22-9
0.8540g/mL
79°C
94% min. (GC)
C10H20O
1.4550 to 1.4580
MFCD00002935
0.854
citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol
QMVPMAAFGQKVCJ-UHFFFAOYSA-N
3,7-dimethyloct-6-en-1-ol
8842
156.27
Liquid
95%
Colorless
222.0°C
Authentic
Glass bottle
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
100 mL
01, 451
15, 2329
Solubility in water: slightly soluble
CC(CCC=C(C)C)CCO
156.27
CHEBI:50462
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/ey

GHS Signal Word: Warning

EINECSNumber : 203-375-0

RTECSNumber : RH3400000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions