missing translation for 'onlineSavingsMsg'
Learn More

Citrisolv Clearing Agent
GREENER_CHOICE

Catalog No. 22143975
Change view
Click to view available options
Quantity:
1 gal.
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
22143975 1 gal. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22143975 Supplier Decon Laboratories Supplier No. DC1601
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Physical Form Liquid
Chemical Name or Material CitriSolv™ Clearing Agent
CAS 5989-27-5, 25013-16-5
CAS Min % 97%, 0.2%
CAS Max % 98%, 0.24%
Packaging Poly Bottle
Color Clear
Quantity 1 gal.
Molecular Weight (g/mol) 46.07 g/mol
Percent Purity 97%
Grade Laboratory
Show More Show Less