missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, trisodium salt dihydrate, ACS reagent

Catalog No. p-4380079
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446332500 250 g Plastic Bottle
AC446330010 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446332500 Supplier Thermo Scientific Chemicals Supplier No. 446332500
Only null left

CAS: 6132-04-3 | C6H5Na3O7·2H2O | 294.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H5Na3O7·2H2O
Molecular Weight (g/mol) 294.09
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Packaging Plastic Bottle
Linear Formula NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O
Quantity 250 g
Merck Index 15,8737
Solubility Information Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol
Formula Weight 294.09
Grade ACS Reagent
Insoluble Matter 0.005% max
Chemical Name or Material Citric Acid, Trisodium Salt Dihydrate

RUO – Research Use Only