Learn More
Citric acid, trisodium salt dihydrate, 99%, pure
CAS: 1545801 | C6H9Na3O9 | 294.1 g/mol
Supplier: Thermo Scientific Chemicals 227130051
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Citric acid, trisodium salt dihydrate | |
| >230°C | |
| White | |
| Authentic | |
| Plastic bottle | |
| NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O | |
| 5 kg | |
| trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
| NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
| trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate | |
| 71474 | |
| 294.09 | |
| Pure |
| 1545801 | |
| >300.0°C | |
| 1.76g/mL | |
| 0.99 | |
| C6H9Na3O9 | |
| MFCD00150031 | |
| 15, 8737 | |
| Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol | |
| C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] | |
| 294.1 | |
| CHEBI:32142 | |
| 0.99 | |
| Crystalline Powder or Crystals |
Chemical Identifiers
| 1545801 | |
| 294.1 | |
| NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
| 71474 | |
| trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate |
| C6H9Na3O9 | |
| MFCD00150031 | |
| trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
| CHEBI:32142 | |
| C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] |
Safety and Handling
Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated.
NFPA
Health:
Signal Word: CAUTION!
RUO – Research Use Only