missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, trisodium salt dihydrate, 99%, for biochemistry, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 447290010

Quantity 1kg
Packaging Plastic bottle

 View more versions of this product

Catalog No. AC447290010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6132-04-3
294.10
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
CHEBI:32142
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
C6H9Na3O9
NLJMYIDDQXHKNR-UHFFFAOYSA-K
71474
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate

Specifications

Citric acid, trisodium salt dihydrate
6132-04-3
C6H9Na3O9
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
71474
294.09
Biochemical
Plastic bottle
(0.1M, water), (0.1M, water) at 0nm, 0.01 max. at 260nm, 0.01 max. at 280nm
99%
NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O
Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
294.10
CHEBI:32142
99%
1kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents