missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Stock Solution, 10% w/w Recommended Storage: 2°C to 8°C (36°F to 46°F)., Reagents

Catalog No. p-200021661
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335598 1 L
01335597 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335598 Supplier Labchem Inc Supplier No. CC2046001A
Only null left

Citric Acid Stock Solution, 10% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Citric Acid Stock Solution, 10% w/w
pH <2
CAS 7732-18-5, 77-92-9
Boiling Point 100°C
Color Colorless
Molecular Formula C6H8O7/H2O
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
Percent Purity 10%
Grade Reagent
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.042
Show More Show Less