missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid Stock Solution, 10% w/w Recommended Storage: 2°C to 8°C (36°F to 46°F)., Reagents
Specifications
Specifications
| Chemical Name or Material | Citric Acid Stock Solution, 10% w/w |
| pH | <2 |
| CAS | 7732-18-5, 77-92-9 |
| Boiling Point | 100°C |
| Color | Colorless |
| Molecular Formula | C6H8O7/H2O |
| Synonym | 2-hydroxypropane-1,2,3-tricarboxylic Acid |
| Percent Purity | 10% |
| Grade | Reagent |
| Quantity | 1 L |
| Show More |