missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Standard, 0.25% (w/w), Reagents

Catalog No. 01335595
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335595 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335595 Supplier Labchem Inc Supplier No. CC204800250C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Citric Acid Standard, 0.25% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Citric Acid Standard, 0.25% (w/w)
pH 2.6
CAS 7732-18-5, 77-92-9
Boiling Point 100°C
Color Colorless
Quantity 250 mL
Packaging Glass Bottle
Physical Form Liquid
Molecular Formula C6H8O7/H2O
Specific Gravity 1.001
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
Percent Purity 0.25%
Grade Reagent
Show More Show Less