missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid, disodium salt sesquihydrate, 99%, pure
CAS: 1545832 | C12H18Na4O17 | 526.22 g/mol
Supplier: Thermo Scientific Chemicals 250240010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Citric acid, disodium salt sesquihydrate | |
| 1545832 | |
| White | |
| Authentic | |
| Plastic bottle | |
| 1·5H2O | |
| 1 kg | |
| unii-cz1032cekr, cz1032cekr, sodium citrate dibasic sesquihydrate, disodium citrate sesquihydrate, disodium hydrogen citrate 1.5 h2o, sodium hydrogen citrate sesquihydrate, citric acid, disodium salt, sesquihydrate, tetrasodium, 3-carboxy-3-hydroxypentanedioate, trihydrate, sodium citrate dibasic sesquihydrate, purum p.a t, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, disodium salt, hydrate 2:3 | |
| HGPVLOQNBSHYEI-UHFFFAOYNA-J | |
| tetrasodium;2-(carboxymethyl)-2-hydroxybutanedioate;trihydrate | |
| 117063816 | |
| 0.99 | |
| Powder |
| Pure | |
| 149.0°C | |
| 4.9 to 5.2 (3% soln.) | |
| 0.99 | |
| C12H18Na4O17 | |
| MFCD00150445 | |
| 14, 2326 | |
| Solubility in water: 500g/L (20°C). | |
| C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Na+].[Na+].[Na+].[Na+] | |
| 526.22 | |
| 263.1 | |
| Pure |
Chemical Identifiers
| 1545832 | |
| 526.22 | |
| HGPVLOQNBSHYEI-UHFFFAOYNA-J | |
| 117063816 | |
| C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Na+].[Na+].[Na+].[Na+] |
| C12H18Na4O17 | |
| MFCD00150445 | |
| unii-cz1032cekr, cz1032cekr, sodium citrate dibasic sesquihydrate, disodium citrate sesquihydrate, disodium hydrogen citrate 1.5 h2o, sodium hydrogen citrate sesquihydrate, citric acid, disodium salt, sesquihydrate, tetrasodium, 3-carboxy-3-hydroxypentanedioate, trihydrate, sodium citrate dibasic sesquihydrate, purum p.a t, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, disodium salt, hydrate 2:3 | |
| tetrasodium;2-(carboxymethyl)-2-hydroxybutanedioate;trihydrate |
RUO – Research Use Only