missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Certified, 10.0% (w/v) ±0.5% (w/v), LabChem™

Supplier:  LabChem LC131502

 View more versions of this product

Catalog No. LC131502

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Citric Acid
90,10
C6H8O7
MFCD00011669
Soluble in water
OC(=O)CC(O)(CC(O)=O)C(O)=O
192.12
CHEBI:30769
10.0% ±0.5% (w/v)
1 L
Liquid
77-92-9,7732-18-5
Colorless
C6H8O7
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
311
192.12
Certified
Poly Bottle
Traceable to NIST

Chemical Identifiers

77-92-9
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
311
2-hydroxypropane-1,2,3-tricarboxylic acid
C6H8O7
MFCD00011669
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
CHEBI:30769
OC(=O)CC(O)(CC(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions