missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Anhydrous, Ultrapure, 500 g, Innovating Science™

Catalog No. S000969
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000969 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000969 Supplier Aldon Corporation Supplier No. IS29037
  • Appearance (Color): Colorless or White
  • Appearance (Form): Crystals or Crystalline Powder
  • Purity: ≥ 99.5%
  • Identification (FTIR): Conforms to Structure
  • Water (KFV): ≤ 0.3%
  • Lead: ≤ 5 ppm
  • Iron: ≤ 5 ppm
  • Oxalate: ≤ 100 ppm
  • Sulfate: ≤ 150 ppm
  • Chloride: ≤ 50 ppm
  • Barium: Passes Test
  • Residue on Ignition: ≤ 0.05%
  • Readily Carbonizable Substances: Passes Test
  • Bacterial Endotoxins: ≤ 0.5 IU/mg
  • DNase: None Detected
  • RNase: None Detected

Chemical Identifiers

CAS 77-92-9
Molecular Weight (g/mol) 192.13

Specifications

Percent Purity 100%
Grade Ultra Pure
Quantity 500 g
Packaging HDPE
Chemical Name or Material Citric acid, anhydrous