missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, Anhydrous Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015531
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015531 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015531 Supplier Merck Emd Millipore Supplier No. PHR10711G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Citric acid is a common ingredient in pharmaceutical formulations. It finds use in antacids and dentrifices because of the effervescent effect it generates upon combination with carbonates or bicarbonates. Citric acid may also be used as a buffering agent and it assists in the dispersion of suspensions in order to ensure the stability of the active ingredients and enhance the efficacy of antioxidants. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 77-92-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HOC(COOH)(CH2 COOH)2
MDL Number MFCD00011669
Melting Point 153°C to 159°C
Synonym Citric acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 192.12
Grade Certified Reference Material
Show More Show Less