missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, Anhydrous, Granular, FCC, 99.5-100.5%, Spectrum™ Chemical
C1281, 77-92-9, C6H8O7
Supplier: Spectrum Chemical Mfg Cor C128112KG
Description
Spectrum™ Chemical Citric Acid, Anhydrous, Granular, FCC is the most widely used organic acid and pH-control agent in foods and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 77-92-9 | |
| C6H8O7 | |
| KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid | |
| 99.5 to 100.5% | |
| 12 kg | |
| Poly Pail |
| 1 | |
| MFCD00011669 | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O | |
| 192.12 | |
| FCC | |
| 0.0005 |
Chemical Identifiers
| 77-92-9 | |
| 192.12 | |
| KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O |
| C6H8O7 | |
| MFCD00011669 | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid |