missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Anhydrous (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-92702
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A940500 500 g Poly Bottle
A9401 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A940500 Supplier Thermo Fisher Scientific Supplier No. FLA940500
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

pH 1.7
Melting Point 153°C
Color White
Quantity 500 g
Ignition Residue 0.02% max.
Packaging Poly Bottle
Physical Form Solid
Assay Percent Range ≥99.5 %
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H
Formula Weight 192.13
Percent Purity ≥99.5%
Grade Certified ACS
Chemical Name or Material Citric Acid Anhydrous
Show More Show Less