missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Anhydrous, ACS Reagent, Reagents

Catalog No. p-200021058
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338179 2.5 kg
01338178 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01338179 Supplier Labchem Inc Supplier No. C11045502.5A5
Only null left

Citric Acid Anhydrous, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Melting Point 153°C
Boiling Point 175°C
Color Colorless/White
Grade ACS
Quantity 2.5 kg
Packaging Poly Bottle
Physical Form Solid
Specific Gravity 1.665
Chemical Name or Material Citric Acid Anhydrous, ACS Reagent