missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Anhydrous, ACS Reagent, Reagents

Catalog No. 01338179
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338179 2.5 kg
01338178 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01338179 Supplier Labchem Inc Supplier No. C11045502.5A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Citric Acid Anhydrous, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Melting Point 153°C
Boiling Point 175°C
Color Colorless/White
Quantity 2.5 kg
Packaging Poly Bottle
Physical Form Solid
Specific Gravity 1.665
Grade ACS
Chemical Name or Material Citric Acid Anhydrous, ACS Reagent