missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid, anhydrous, ACS, 99.5+%
Description
Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 153°C to 154°C |
| Assay Percent Range | >99.5+% |
| Beilstein | 782061 |
| Solubility Information | It is very soluble in water,freely soluble in ethanol (96 %) and sparingly soluble in ether. |
| Formula Weight | 192.13 |
| Grade | ACS Reagent |
| Density | 1.665 g/mL |
| Quantity | 100 g |
| Physical Form | Crystalline |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H319-H335-H500 |
| Precautionary Statement | P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c |
| EINECSNumber | 201-069-1 |
| RTECSNumber | GE7350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only