missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Anhydrous, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18609809
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609809 25 g Poly Bottle
18609810 125 g Poly Bottle
18609813 500 g Poly Bottle
18609814 2.5 kg Poly Pail
18609811 12 kg Poly Pail
18609812 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609809 Supplier Spectrum Chemical Mfg Cor Supplier No. C128025GM
Only null left

Chemical Identifiers

CAS 77-92-9

Specifications

CAS Max % 1
Percent Purity 99.5%
Grade ACS
Quantity 25 g
Packaging Poly Bottle
Residue after Ignition 0.0002

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.