missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Anhydrous, 99.5 to 100.5%, ACS, MilliporeSigma™

Catalog No. MCX17233
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCX17233 2.5 kg
MCX17231 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MCX17233 Supplier Technidata Supplier No. CX17233

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Color White
Density 560 kg/m3
Quantity 2.5 kg
Ignition Point 345°C
Physical Form Crystalline Solid
Assay Percent Range 99.5 to 100.5%
Vapor Pressure < - 0.1 hPa (20°C)
Formula Weight 192.12
Chemical Name or Material Citric Acid, Anhydrous, For Synthesis
Recommended Storage Store at +5°C to +30°C