missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, 99+%

Catalog No. AAA1039530
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1039530 250 g
AAA103950B 1000 g
AAA103950I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1039530 Supplier Thermo Scientific Chemicals Supplier No. A1039530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.

Solubility
Soluble in water, alcohol, ether, dimethylsufoxide and ethylacetate.

Notes
Hygroscopic. Incompatible with alkalies, bases, metal nitrates and oxidizing agents.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

pH 1.6
Melting Point 153°C to 154°C
Density 1.665
Quantity 250 g
Odor Odorless
Beilstein 782061
Merck Index 14,2326
Solubility Information Soluble in water,alcohol,ether,dimethylsufoxide and ethylacetate.
Formula Weight 192.13
Percent Purity ≥99%
Chemical Name or Material Citric acid
Show More Show Less
Hazard Category H319-H335-H500
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 201-069-1
RTECSNumber GE7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only