missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, 99.5 to 100.5% (based on Anhydrous substance), Honeywell Fluka™

Catalog No. 60045753
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
100 g
2.5 kg
6 x 1 kg
6 x 2.5 kg
Catalog No. Quantity
60045753 6 x 1 kg
6002241 100 g
6002242 1 kg
6002243 2.5 kg
60045754 6 x 2.5 kg
5 options

Catalog No. 60045753

Supplier: Honeywell Chemicals 271096X1KG

Only null left

Meets analytical specification of BP, Ph. Eur., USP, E330, Anhydrous, 99.5-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Linear Formula HOC(COOH)(CH2COOH)2
UN Number NONH for all modes of transport
Solubility Information Soluble in water, 383g/L at 25°C, Sparingly soluble in Diethyl Ether, Freely soluble in alcohol
Formula Weight 192.12
Percent Purity 99.5 to 100.5% (based on anhydrous substance)
Quantity 6 x 1 kg
Explosion Limit 8% (65°F)
Residue after Ignition ≤0.05% (as SO4)
Chemical Name or Material Citric Acid, Meets analytical specification of Ph. Eur.BP, USP
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 201-069-1
RTECSNumber GE7350000