missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Citrate-buffered saline (20X)

Catalog No. AAJ63666K3
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63666K3 2 L
AAJ63666K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63666K3 Supplier Thermo Scientific™ Supplier No. J63666K3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 4-3-6132
Concentration 20X
Physical Form Liquid
Quantity 2 L
Synonym SSC buffer, 20X; Saline-sodium citrate buffer, 20X
Molecular Weight (g/mol) 294.10