missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Citrate-buffered saline (20X)

Catalog No. AAJ63666K2
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63666K2 1 L
AAJ63666K3 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63666K2 Supplier Thermo Scientific™ Supplier No. J63666K2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 4-3-6132
Concentration 20X
Physical Form Liquid
Quantity 1 L
Synonym SSC buffer, 20X; Saline-sodium citrate buffer, 20X
Molecular Weight (g/mol) 294.10