Learn More
Electron Microscopy Sciences Citrate Buffer | pH 6.0 | 100ml
Catalog No. 501926525
Encompass
This buffer is intended for heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. The use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens.
Catalog No. 501926525 Supplier Electron Microscopy Sciences Supplier No. 6414207
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This buffer is intended for heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. The use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens.