missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 4.5

Catalog No. AAJ60024AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60024AK 250 mL
AAJ60024AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60024AK Supplier Thermo Scientific Chemicals Supplier No. J60024AK

CAS: 6132-04-3294.10 g/mol

Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.

Solubility
Fully miscible with water. Insoluble in ethanol.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 4-3-6132
Molecular Weight (g/mol) 294.10

Specifications

Color Colorless
Concentration 0.5M buffer soln.
pH 4.5
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Recommended Storage Keep cold

RUO – Research Use Only