missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 3.0

CAS: 6132-04-3294.10 g/mol

Supplier:  Thermo Scientific Chemicals J61391AK

 View more versions of this product

Catalog No. AAJ61391AK


Only null left
Explore available promotions

Description

Description

Citrate is an intermediate in the TCA cycle and fatty acid synthesis. It is an allosteric modulator of acetyl-CoA carboxylase, the enzyme that regulates the conversion of acetyl-CoA to malonyl-CoA. It has anticoagulant activity and is a calcium chelator. Its function is to break protein cross-links; thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citrate is an intermediate in the TCA cycle and fatty acid synthesis. It is an allosteric modulator of acetyl-CoA carboxylase, the enzyme that regulates the conversion of acetyl-CoA to malonyl-CoA. It has anticoagulant activity and is a calcium chelator. Its function is to break protein cross-links; thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies.

Solubility
Fully miscible in water. Insoluble in ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

4-3-6132
0.5M buffer soln.
Liquid
Odorless
Colorless
3.0
250 mL
294.10

Chemical Identifiers

4-3-6132
294.10
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only