missing translation for 'onlineSavingsMsg'
Learn More

Citral Dimethyl Acetal (cis- and trans- mixture) 90.0+%, TCI America™

Catalog No. C121125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C121125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C121125ML Supplier TCI America Supplier No. C121125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7549-37-3
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00008909
InChI Key ZSKAJFSSXURRGL-PKNBQFBNSA-N
Synonym 1,1-Dimethoxy-3,7-dimethyl-2,6-octadiene
PubChem CID 5352435
IUPAC Name (2E)-1,1-dimethoxy-3,7-dimethylocta-2,6-diene
SMILES CC(=CCCC(=CC(OC)OC)C)C

Spécifications

Color Yellow
Boiling Point 103°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Citral Dimethyl Acetal (cis- and trans- mixture)
RTECSNumber RG5330000
TSCA Yes