missing translation for 'onlineSavingsMsg'
Learn More

Citral, cis + trans, 95%

Catalog No. AAA1616936
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1616936 500 g
AAA1616918 50 g
AAA1616922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1616936 Supplier Thermo Scientific Chemicals Supplier No. A1616936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5392-40-5 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5392-40-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00006997
InChI Key WTEVQBCEXWBHNA-JXMROGBWSA-N
Synonym citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
PubChem CID 638011
ChEBI CHEBI:16980
IUPAC Name (2E)-3,7-dimethylocta-2,6-dienal
SMILES CC(C)=CCC\C(C)=C\C=O

Specifications

Melting Point <-10°C
Density 0.888
Boiling Point 228°C to 229°C
Flash Point 98°C (208°F)
Refractive Index 1.488
Quantity 500 g
Beilstein 1721871
Merck Index 14,2322
Formula Weight 152.24
Percent Purity 95%
Chemical Name or Material Citral, cis + trans
Show More Show Less
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H315-H317
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
EINECSNumber 226-394-6
RTECSNumber RG5075000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only