missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citraconic Anhydride 98.0+%, TCI America™
Supplier: TCI America M0365100G
Specifications
| Citraconic Anhydride | |
| Yellow | |
| C5H4O3 | |
| 100 g | |
| citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx | |
| CC1=CC(=O)OC1=O | |
| 112.084 | |
| 112.08 | |
| Liquid |
| 616-02-4 | |
| 215°C | |
| MFCD00005522 | |
| 2810 | |
| AYKYXWQEBUNJCN-UHFFFAOYSA-N | |
| 3-methylfuran-2,5-dione | |
| 12012 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 616-02-4 | |
| 112.084 | |
| AYKYXWQEBUNJCN-UHFFFAOYSA-N | |
| 12012 | |
| CC1=CC(=O)OC1=O |
| C5H4O3 | |
| MFCD00005522 | |
| citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx | |
| 3-methylfuran-2,5-dione |
Safety and Handling
EINECSNumber : (5)-6177
RTECSNumber : GE6825000
TSCA : Yes