missing translation for 'onlineSavingsMsg'
Learn More

Citraconic Anhydride 98.0+%, TCI America™

Catalog No. M036525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M036525G 25 g
M0365100G 100 g
M0365500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M036525G Supplier TCI America Supplier No. M036525G

Chemical Identifiers

CAS 616-02-4
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.084
MDL Number MFCD00005522
InChI Key AYKYXWQEBUNJCN-UHFFFAOYSA-N
Synonym citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx
PubChem CID 12012
IUPAC Name 3-methylfuran-2,5-dione
SMILES CC1=CC(=O)OC1=O

Specifications

Color Yellow
Boiling Point 215°C
Quantity 25 g
UN Number 2810
Formula Weight 112.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Citraconic Anhydride
EINECSNumber (5)-6177
RTECSNumber GE6825000
TSCA Yes