missing translation for 'onlineSavingsMsg'
Learn More

Citraconic anhydride, 98%

Catalog No. p-7044202
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
AAL0523814 25 g
AAL0523822 100 g
AAL0523836 500 g
3 options

Catalog No. AAL0523814

Supplier: Thermo Scientific Chemicals L0523814

Only null left

CAS: 616-02-4 | C5H4O3 | 112.084 g/mol

Citraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.

Solubility
Miscible with water, alcohol, acetone and ether.

Notes
Moisture sensitive. Incompatible with bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 616-02-4
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.084
MDL Number MFCD00005522
InChI Key AYKYXWQEBUNJCN-UHFFFAOYSA-N
Synonym citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx
PubChem CID 12012
IUPAC Name 3-methylfuran-2,5-dione
SMILES CC1=CC(=O)OC1=O

Specifications

Melting Point 7°C to 8°C
Density 1.243
Boiling Point 213°C to 214°C
Flash Point 101°C (213°F)
Refractive Index 1.471
Quantity 25 g
UN Number UN2810
Beilstein 1835
Sensitivity Moisture sensitive
Solubility Information Miscible with water,alcohol,acetone and ether.
Formula Weight 112.08
Percent Purity 98%
Chemical Name or Material Citraconic anhydride
Show More Show Less
Hazard Category H311-H314-H317-H334
Hazard Statement GHS H Statement
H311-H315-H319
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 210-459-0
RTECSNumber GE6825000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only