missing translation for 'onlineSavingsMsg'
Learn More

Citraconic anhydride, 98%

Catalog No. p-93205
Change view
Click to view available options
Quantity:
100 g
25 g
250 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC154790250 25 g Glass bottle
AC154791000 100 g Glass bottle
AC154792500 250 g Glass bottle
3 options

Catalog No. AC154790250

Supplier: Thermo Scientific Chemicals 154790250

Only null left

CAS: 616-02-4 | C5H4O3 | 112.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-02-4
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00005522
InChI Key AYKYXWQEBUNJCN-UHFFFAOYSA-N
Synonym citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx
PubChem CID 12012
IUPAC Name 3-methylfuran-2,5-dione
SMILES CC1=CC(=O)OC1=O

Specifications

Melting Point 6°C to 7°C
Color Colorless to Yellow
Density 1.2450g/mL
Boiling Point 213°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4700 to 1.472
Quantity 25 g
Beilstein 17, 440
Specific Gravity 1.245
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in ethanol and ether
Formula Weight 112.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Citraconic anhydride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-459-0
RTECSNumber GE6825000
TSCA TSCA

RUO – Research Use Only