missing translation for 'onlineSavingsMsg'
Learn More

Citraconic Acid 98.0+%, TCI America™

Catalog No. C0363500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0363500G 500 g
C036325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0363500G Supplier TCI America Supplier No. C0363500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 498-23-7
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.10
MDL Number MFCD00078086
InChI Key HNEGQIOMVPPMNR-IHWYPQMZSA-N
Synonym 2-methylmaleate, citraconate 2-, z-2-methylbut-2-enedioate
PubChem CID 5461090
ChEBI CHEBI:30719
IUPAC Name (2Z)-2-methylbut-2-enedioic acid
SMILES C\C(=C\C(O)=O)C(O)=O

Specifications

Melting Point 87°C
Color White
Quantity 500 g
Formula Weight 130.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Citraconic Acid
EINECSNumber (2)-3387
RTECSNumber GE6650000
TSCA No