missing translation for 'onlineSavingsMsg'
Learn More

Cistus Oil, Spectrum™ Chemical

$293.07 - $2177.71

Chemical Identifiers

CAS 8016-26-0
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609803
View Documents
Spectrum Chemical Mfg Cor
C18425ML
5 mL Amber Glass Bottle
Each for $293.07
Only null left
 
18609802
View Documents
Spectrum Chemical Mfg Cor
C184225ML
25 mL Amber Glass Bottle
Each for $871.00
Only null left
 
18609801
View Documents
Spectrum Chemical Mfg Cor
C1842100ML
100 mL Amber Glass Bottle
Each for $2,177.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

8016-26-0

Specifications

Liquid
1
Light Brown
Ungraded
8016-26-0
Amber Glass Bottle
5 mL
Cistus Oil
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.