missing translation for 'onlineSavingsMsg'
Learn More

cis-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 97.0+%, TCI America™

Catalog No. B43311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43311G 1 g
B43315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43311G Supplier TCI America Supplier No. B43311G
Only null left

Chemical Identifiers

CAS 184954-75-4
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD07367780
InChI Key AKVIZYGPJIWKOS-DTWKUNHWSA-N
Synonym cis-N1-Boc-1,2-cyclohexanediamine, cis-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, cis-2-Amino-1-(Boc-amino)cyclohexane
PubChem CID 1514392
IUPAC Name tert-butyl N-[(1R,2S)-2-aminocyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCCC1N

Specifications

Melting Point 46°C
Color White
Quantity 1 g
Formula Weight 214.31
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material cis-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine
TSCA No
Recommended Storage Refrigerator