missing translation for 'onlineSavingsMsg'
Learn More

cis-Jasmone 92.0+%, TCI America™

Catalog No. p-7131732
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
J00035ML 5 mL
J000325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. J00035ML Supplier TCI America Supplier No. J00035ML
Only null left

Chemical Identifiers

CAS 488-10-8
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00001402,MFCD00001402,MFCD00001402
InChI Key XMLSXPIVAXONDL-PLNGDYQASA-N
Synonym trans-jasmone, e-jasmone, unii-7tcs3y45dr, 7tcs3y45dr, 3-methyl-2-e-pent-2-enyl cyclopent-2-en-1-one, 2-cyclopenten-1-one, 3-methyl-2-2-pentenyl-, e, 3-methyl-2-pent-2-en-1-yl cyclopent-2-en-1-one, 3-methyl-2-2e-pent-2-en-1-yl cyclopent-2-en-1-one, jasmone, e, 3-methyl-2-2-pentenyl-2-cyclopenten-1-one
PubChem CID 1549019
IUPAC Name 3-methyl-2-[(2Z)-pent-2-en-1-yl]cyclopent-2-en-1-one
SMILES CC\C=C/CC1=C(C)CCC1=O

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 mL
Formula Weight 164.25
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-Jasmone
EINECSNumber (3)-2394
RTECSNumber GY7301000
TSCA Yes