missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-Jasmone 92.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 135°C |
| Quantity | 5 mL |
| Formula Weight | 164.25 |
| Percent Purity | ≥92.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | cis-Jasmone |
Safety and Handling
| EINECSNumber | (3)-2394 |
| RTECSNumber | GY7301000 |
| TSCA | Yes |