missing translation for 'onlineSavingsMsg'
Learn More

cis-Epoxysuccinic Acid 97.0+%, TCI America™

Catalog No. p-7129002
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04495G 5 g
E044925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04495G Supplier TCI America Supplier No. E04495G
Only null left

Chemical Identifiers

CAS 16533-72-5
Molecular Formula C4H4O5
Molecular Weight (g/mol) 132.071
MDL Number MFCD00191645
InChI Key DCEMCPAKSGRHCN-XIXRPRMCSA-N
Synonym cis-epoxysuccinic acid, 2r,3s-oxirane-2,3-dicarboxylic acid, cis-oxirane-2,3-dicarboxylic acid, 2,3-oxiranedicarboxylic acid, 2r,3s-rel, 2,3-oxiranedicarboxylic acid, 2r,3s-rel-9ci
PubChem CID 2734802
IUPAC Name (2S,3R)-oxirane-2,3-dicarboxylic acid
SMILES C1(C(O1)C(=O)O)C(=O)O

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Formula Weight 132.07
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-Epoxysuccinic Acid