missing translation for 'onlineSavingsMsg'
Learn More

cis-Epoxysuccinic acid, 97%

Catalog No. p-3212533
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC372640050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC372640050 Supplier Thermo Scientific Chemicals Supplier No. 372640050

CAS: 16533-72-5 | C4H4O5 | 132.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16533-72-5
Molecular Formula C4H4O5
Molecular Weight (g/mol) 132.07
MDL Number MFCD00191645
InChI Key DCEMCPAKSGRHCN-XIXRPRMCSA-N
Synonym cis-epoxysuccinic acid, 2r,3s-oxirane-2,3-dicarboxylic acid, cis-oxirane-2,3-dicarboxylic acid, 2,3-oxiranedicarboxylic acid, 2r,3s-rel, 2,3-oxiranedicarboxylic acid, 2r,3s-rel-9ci
PubChem CID 2734802
IUPAC Name (2S,3R)-oxirane-2,3-dicarboxylic acid
SMILES C1(C(O1)C(=O)O)C(=O)O

Specifications

Melting Point 133°C to 135°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 132.07
Percent Purity 97%
Chemical Name or Material cis-Epoxysuccinic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 240-604-3

RUO – Research Use Only