missing translation for 'onlineSavingsMsg'
Learn More

cis-Diammineplatinum(II) Dichloride, TCI America™

$91.33 - $428.73

Chemical Identifiers

CAS 15663-27-1
Molecular Formula Cl2H6N2Pt
Molecular Weight (g/mol) 300.04
MDL Number MFCD00011623
InChI Key BSJGASKRWFKGMV-UHFFFAOYSA-L
Synonym Cisplatin, cis-Diamminedichloroplatinum(II), cis-Platinum(II) Diammine Dichloride
IUPAC Name dichloroplatinumbis(ylium) diamine
SMILES N.N.Cl[Pt++]Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D3371100MG
View Documents
TCI America
D3371100MG
100 mg
Each for $91.33
Only null left
 
D33711G
View Documents
TCI America
D33711G
1 g
Each for $428.73
Only null left
 
Description

Description

Specifications

Chemical Identifiers

15663-27-1
300.04
BSJGASKRWFKGMV-UHFFFAOYSA-L
dichloroplatinumbis(ylium) diamine
Cl2H6N2Pt
MFCD00011623
Cisplatin, cis-Diamminedichloroplatinum(II), cis-Platinum(II) Diammine Dichloride
N.N.Cl[Pt++]Cl

Specifications

15663-27-1
Yellow
MFCD00011623
3288
BSJGASKRWFKGMV-UHFFFAOYSA-L
dichloroplatinumbis(ylium) diamine
300.05
cis-Diammineplatinum(II) Dichloride
270°C
Cl2H6N2Pt
100 mg
Cisplatin, cis-Diamminedichloroplatinum(II), cis-Platinum(II) Diammine Dichloride
N.N.Cl[Pt++]Cl
300.04
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TP2450000

TSCA : Yes

SDS
Documents

Documents