Learn More
cis,cis-1,5-Cyclooctadiene, 99%, stab. with 50-200ppm Irganox 1076
CAS: 1552-12-1 | C8H12 | 108.184 g/mol
$62.14 - $174.60
Chemical Identifiers
| CAS | 1552-12-1 |
|---|---|
| Molecular Formula | C8H12 |
| Molecular Weight (g/mol) | 108.184 |
| MDL Number | MFCD00001752 |
| InChI Key | VYXHVRARDIDEHS-QGTKBVGQSA-N |
| Synonym | 1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod |
| PubChem CID | 82916 |
| IUPAC Name | (1Z,5Z)-cycloocta-1,5-diene |
| SMILES | C1CC=CCCC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB21196AK
|
Thermo Scientific Chemicals
B21196AK |
250 mL |
Each for $62.14
|
|
|||||
|
AAB21196AU
|
Thermo Scientific Chemicals
B21196AU |
1000 mL |
Each for $174.60
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1552-12-1 | |
| 108.184 | |
| VYXHVRARDIDEHS-QGTKBVGQSA-N | |
| 82916 | |
| C1CC=CCCC=C1 |
| C8H12 | |
| MFCD00001752 | |
| 1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod | |
| (1Z,5Z)-cycloocta-1,5-diene |
Specifications
| 1552-12-1 | |
| 0.880 | |
| 32°C (89°F) | |
| C8H12 | |
| MFCD00001752 | |
| UN2520 | |
| Air Sensitive | |
| VYXHVRARDIDEHS-QGTKBVGQSA-N | |
| (1Z,5Z)-cycloocta-1,5-diene | |
| 82916 | |
| 99% | |
| cis, cis-1, 5-Cyclooctadiene, Stab. With 50-200ppm Irganox 1076 |
| °C to 70°C | |
| 151°C to 152°C | |
| 0.99 | |
| 1.494 | |
| 250 mL | |
| 1209288 | |
| 1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod | |
| C1CC=CCCC=C1 | |
| 108.184 | |
| 108.18 | |
| Liquid |
Safety and Handling
GHS H Statement
H226-H302-H332-H315-H319-H317-H335
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c
H226-H302+H332-H315-H317-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOOCTADIENES
EINECSNumber : 216-291-4
RTECSNumber : GX9620000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only